Learn More
Sino Biological Anti-PCSK9 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80005MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat PCSK9 / NARC1 (rR PCSK9 / NARC1; Catalog#80005-R08H; NP_954862.2; Met1-Gln691). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-2889