Learn More
Sino Biological Recombinant Anti-PCSK9 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80005R00720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat PCSK9 / NARC1 (rR PCSK9 / NARC1; Catalog#80005-R08H; NP_954862.2; Met1-Gln691).
Catalog No. 50-255-2886