Learn More
Sino Biological Anti-PCSK9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80005RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat PCSK9 (rR PCSK9; Catalog#80005-R08H; NP_954862.2; Met 1-Gln 691). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2884