Learn More
Sino Biological Rat PDCD1LG2 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80454R08H100
Encompass_Preferred
A DNA sequence encoding the rat PDCD1LG2 (NP_001101052.2) (Met1-Arg219) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2002