Learn More
Sino Biological Anti-PDGF alpha receptor/PDGFRA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80433T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat PDGFRA / CD140a (rR PDGFRA / CD140a; Catalog#80433-R08H; NP_036934.1; Met1-Glu523). PDGFRA / CD140a specific IgG was purified by Rat PDGFRA / CD140a affinity chromatography.
Catalog No. 50-241-0867