Learn More
Sino Biological Rat PDGFRA Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80433R02H100
Encompass_Preferred
A DNA sequence encoding the rat PDGFRA (G3V6A0) (Met1-Glu523) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1964