Learn More
Sino Biological Rat PDPN Protein 200ug (Sino Biological)
Supplier:  Sino Biological 81154R02H200
Encompass_Preferred
A DNA sequence encoding the rat Pdpn (NP_062231.1) (Met1-Leu135) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1656