Learn More
Biomatik Corporation Rat Pepsin (PP) ELISA Kit
Supplier:  Biomatik Corporation EKU10192
Encompass_Preferred
Rat Pepsin (PP) ELISA Kit for detecting PP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.90ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8362