Learn More
Biomatik Corporation Rat Perilipin 1 (PLIN1) ELISA Kit
Supplier:  Biomatik Corporation EKU11330
Encompass_Preferred
Rat Perilipin 1 (PLIN1) ELISA Kit for detecting PLIN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43884. Target Synonyms: PERI; PLIN1; Lipid Droplet Associated Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6194