Learn More
Biomatik Corporation Rat Perilipin 3 (PLIN3) ELISA Kit
Supplier:  Biomatik Corporation EKU11331
Encompass_Preferred
Rat Perilipin 3 (PLIN3) ELISA Kit for detecting PLIN3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A8I6A147. Target Synonyms: M6PRBP1; TIP47; PP17; Mannose-6-Phosphate Receptor Binding Protein 1; Placental Protein 17; 47 kDa mannose 6-phosphate receptor-binding protein; Cargo selection TIP47. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6196