Learn More
Biomatik Corporation Rat Peroxidasin Homolog (PXDN) ELISA Kit
Supplier:  Biomatik Corporation EKU11332
Encompass_Preferred
Rat Peroxidasin Homolog (PXDN) ELISA Kit for detecting PXDN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A0G2KAH2. Target Synonyms: PRG2; PXN; MG50; VPO; VPO1; Melanoma-associated antigen MG50; Vascular peroxidase 1; p53-responsive gene 2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5315