Learn More
American Research Products Inc Rat PeroXisome Proliferator ac
Catalog No. 5000329682
Encompass
Rat PeroXisome Proliferator-actiVated receptor y PPAR-y ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29682 Supplier American Research Products Inc Supplier No. CSBE08624R
Rat PeroXisome Proliferator-actiVated receptor y PPAR-y ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich