Learn More
American Research Products Inc Rat peroXisome proliferators a
Catalog No. 5000328682
Encompass
Rat peroXisome proliferators actiVator receptors alpha PPAR-a ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28682 Supplier American Research Products Inc Supplier No. CSBE09755R
Rat peroXisome proliferators actiVator receptors alpha PPAR-a ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich