Learn More
Sino Biological Anti-Pepsinogen C/PGC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80097RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Pepsinogen C / PGC (Catalog#80097-R08H; NP_579818.1; Met1-Val392). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2832