Learn More
Sino Biological Anti-Pepsinogen C/PGC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80097T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Pepsinogen C / PGC (Catalog#80097-R08H; NP_579818.1; Met1-Val392). Pepsinogen C / PGC specific IgG was purified by Rat Pepsinogen C / PGC affinity chromatography.
Catalog No. 50-255-2831