Learn More
Biomatik Corporation Rat Phosphodiesterase 3A, cGMP Inhibited (PDE3A) ELISA Kit
Supplier:  Biomatik Corporation EKU06590
Encompass_Preferred
Rat Phosphodiesterase 3A, cGMP Inhibited (PDE3A) ELISA Kit for detecting PDE3A in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62865. Gene ID: 50678. Target Synonyms: CGI-PDE; Cyclic GMP-inhibited phosphodiesterase A; cGMP-inhibited 3',5'-cyclic phosphodiesterase A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6640