Learn More
Biomatik Corporation Rat Phosphofructokinase, Muscle (PFKM) ELISA Kit
Supplier:  Biomatik Corporation EKU06600
Encompass_Preferred
Rat Phosphofructokinase, Muscle (PFKM) ELISA Kit for detecting PFKM in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47858. Gene ID: 65152. Target Synonyms: PFK1; PFKX; Phosphofructokinase 1; Phosphofructo-1-kinase isozyme A; 6-Phosphofructokinase Polypeptide X; Fructose-6-Phosphate 1-Phosphotransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8820