Learn More
Biomatik Corporation Rat Phosphoinositide-3-Kinase Catalytic Beta Polypeptide (PIK3Cb) ELISA Kit
Supplier:  Biomatik Corporation EKU12497
Encompass_Preferred
Rat Phosphoinositide-3-Kinase Catalytic Beta Polypeptide (PIK3Cb) ELISA Kit for detecting PIK3Cb in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1L0. Gene ID: 85243. Target Synonyms: PI3K; PI3Kbeta; PIK3C1; p110-BETA; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7593