Learn More
American Research Products Inc Rat Phospholipid hydroperoXide
Catalog No. 5000327326
Encompass
Rat Phospholipid hydroperoXide glutathione peroXidase mitochondrial (GPX4) ELISA kit from CUSABIO Detection Range 4 68 U/mL-300 U/mL Sensitivity 1 17 U/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27326 Supplier American Research Products Inc Supplier No. CSBEL009869RA
Rat Phospholipid hydroperoXide glutathione peroXidase mitochondrial (GPX4) ELISA kit from CUSABIO Detection Range 4 68 U/mL-300 U/mL Sensitivity 1 17 U/mL Sample serum plasma tissue homogenates Method Sandwich