Learn More
BIOHIPPO Rat PICALM Microsample
SDP
Catalog No. 5000314811
Encompass
Rat PICALM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PICALM in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 102 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PICALM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PICALM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PICALM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14811 Supplier BIOHIPPO Supplier No. BHE15210658
Rat PICALM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PICALM in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 102 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PICALM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PICALM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PICALM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color