Learn More
Biomatik Corporation Rat Piccolo (PCLO) ELISA Kit
Supplier:  Biomatik Corporation EKU12418
Encompass_Preferred
Rat Piccolo (PCLO) ELISA Kit for detecting PCLO in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JKS6. Gene ID: 56768. Target Synonyms: ACZ; Aczonin; Presynaptic Cytomatrix Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7723