Learn More
American Research Products Inc Rat pigment epithelium deriVed
Catalog No. 5000329618
Encompass
Rat pigment epithelium-deriVed factor PEDF ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29618 Supplier American Research Products Inc Supplier No. CSBE08819R
Rat pigment epithelium-deriVed factor PEDF ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich