Learn More
Biomatik Corporation Rat Pigment Epithelium Derived Factor (PEDF) CLIA Kit
Supplier:  Biomatik Corporation EKU09812
Encompass_Preferred
Rat Pigment Epithelium Derived Factor (PEDF) CLIA Kit for detecting PEDF in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q80ZA3. Gene ID: 287526. Target Synonyms: SERPINF1; EPC1; PIG35; SDF3; Serpin F1; Cell proliferation-inducing gene 35; Serpin Peptidase Inhibitor,Clade F Member 1 (Alpha-2 Antiplasmin) ; Stromal Cell Derived Factor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4770