Learn More
American Research Products Inc Rat placenta growth factor PL
Catalog No. 5000329808
Encompass
Rat placenta growth factor PLGF ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29808 Supplier American Research Products Inc Supplier No. CSBE07400R
Add to Cart
Add to Cart
Rat placenta growth factor PLGF ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich