Learn More
American Research Products Inc Rat plasmin antiplasmin comple
Catalog No. 5000329619
Encompass
Rat plasmin-antiplasmin compleX PAP ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum tissue homogenates Method Sandwich
Catalog No. 50-003-29619 Supplier American Research Products Inc Supplier No. CSBE08435R
Rat plasmin-antiplasmin compleX PAP ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum tissue homogenates Method Sandwich