Learn More
Biomatik Corporation Rat Plasminogen Activator Inhibitor 2 (PAI2) ELISA Kit
Supplier:  Biomatik Corporation EKU06664
Encompass_Preferred
Rat Plasminogen Activator Inhibitor 2 (PAI2) ELISA Kit for detecting PAI2 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29524. Gene ID: 60325. Target Synonyms: SERPINB2; PLANH2; Serpin Peptidase Inhibitor Clade B Member 2; Monocyte Arg-serpin; Placental plasminogen activator inhibitor; Serpin B2; Urokinase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8817