Learn More
American Research Products Inc Rat plasminogen actiVator inhi
Catalog No. 5000326505
Encompass
Rat plasminogen actiVator inhibitor 1 PAI1 ELISA Kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26505 Supplier American Research Products Inc Supplier No. CSBE07948R
Rat plasminogen actiVator inhibitor 1 PAI1 ELISA Kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich