Learn More
Biomatik Corporation Rat Plasminogen Activator Inhibitor 1 (PAI1) CLIA Kit
Supplier:  Biomatik Corporation EKU09953
Encompass_Preferred
Rat Plasminogen Activator Inhibitor 1 (PAI1) CLIA Kit for detecting PAI1 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P20961. Gene ID: 24617. Target Synonyms: SERPINE1; PLANH1; Serpin Peptidase Inhibitor Clade E Member 1; Nexin,Plasminogen Activator Inhibitor Type 1 Serpin E1; Endothelial plasminogen activator inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5918