Learn More
American Research Products Inc Rat platelet actiVating factor
Catalog No. 5000326528
Encompass
Rat platelet actiVating factor PAF ELISA KIT from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26528 Supplier American Research Products Inc Supplier No. CSBE07930R
Rat platelet actiVating factor PAF ELISA KIT from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich