Learn More
Biomatik Corporation Rat Platelet Derived Growth Factor Receptor Alpha (PDGFRa) ELISA Kit
Supplier:  Biomatik Corporation EKU06692
Encompass_Preferred
Rat Platelet Derived Growth Factor Receptor Alpha (PDGFRa) ELISA Kit for detecting PDGFRa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20786. Gene ID: 25267. Target Synonyms: CD140a; PDGFR2; Rhe-PDGFRA; CD140 antigen-like family member A; Platelet-derived growth factor receptor 2; Alpha-type platelet-derived growth factor receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5178