Learn More
American Research Products Inc Rat Platelet deriVed growth fa
Catalog No. 5000327483
Encompass
Rat Platelet-deriVed growth factor CC (PDGF-CC) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27483 Supplier American Research Products Inc Supplier No. CSBEL017710RA
Rat Platelet-deriVed growth factor CC (PDGF-CC) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich