Learn More
Biomatik Corporation Rat Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit
Supplier:  Biomatik Corporation EKU11343
Encompass_Preferred
Rat Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit for detecting PDGFA in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28576. Gene ID: 25266. Target Synonyms: PDGF-A; PDGF1; Platelet Derived Growth Factor Alpha Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6805