Learn More
American Research Products Inc Rat Platelet DeriVed Growth Fa
Catalog No. 5000326594
Encompass
Rat Platelet-DeriVed Growth Factor AB PDGF-AB ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26594 Supplier American Research Products Inc Supplier No. CSBE04703R
Rat Platelet-DeriVed Growth Factor AB PDGF-AB ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich