Learn More
American Research Products Inc Rat platelet deriVed growth fa
Catalog No. 5000329646
Encompass
Rat platelet-deriVed growth factor-BB PDGF-BB ELISA Kit from CUSABIO Detection Range 39 063 pg/mL-2500 pg/mL Sensitivity 9 77 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29646 Supplier American Research Products Inc Supplier No. CSBE08924R
Rat platelet-deriVed growth factor-BB PDGF-BB ELISA Kit from CUSABIO Detection Range 39 063 pg/mL-2500 pg/mL Sensitivity 9 77 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich