Learn More
American Research Products Inc Rat Platelet DeriVed Growth Fa
Catalog No. 5000328852
Encompass
Rat Platelet-DeriVed Growth Factor A PDGF-A ELISA kit from CUSABIO Detection Range 31 25pg/ml-2000pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28852 Supplier American Research Products Inc Supplier No. CSBE14313R
Rat Platelet-DeriVed Growth Factor A PDGF-A ELISA kit from CUSABIO Detection Range 31 25pg/ml-2000pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich