Learn More
American Research Products Inc Rat platelet membrane glycopro
Catalog No. 5000326718
Encompass
Rat platelet membrane glycoprotein IIbIIIa GP-IIbIIIa ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26718 Supplier American Research Products Inc Supplier No. CSBE08776R
Rat platelet membrane glycoprotein IIbIIIa GP-IIbIIIa ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich