Learn More
Sino Biological Rat PLAU Protein 50ug (Sino Biological)
Supplier:  Sino Biological 81600R08H50
Encompass_Preferred
A DNA sequence encoding the rat Plau (NP_037217.3) (Met1-Phe432) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1820