Learn More
Biomatik Corporation Rat Pleiotrophin (PTN) CLIA Kit
Supplier:  Biomatik Corporation EKU09662
Encompass_Preferred
Rat Pleiotrophin (PTN) CLIA Kit for detecting PTN in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P63090. Gene ID: 24924. Target Synonyms: HARP; HBGF8; HBNF; NEGF1; Heparin Binding Growth Factor 8; Neurite Growth-Promoting Factor 1; Heparin Affin Regulatory Peptide; Heparin Binding Growth Associated Molecule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7785