Learn More
Active Motif Rat Positive Control Primer Set Actb
SDP
Catalog No. NC1850919
Encompass
Model Organism Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with samples from rat, zerbrafish, Drosophila or budding yeast. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP DNA from multiple samples.
Catalog No. NC1850919 Supplier Active Motif Supplier No. 71026
Model Organism Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with samples from rat, zerbrafish, Drosophila or budding yeast. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP DNA from multiple samples.