Learn More
BIOHIPPO Rat PP Microsample
SDP
Catalog No. 5000314088
Encompass
Rat PP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PP in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 8 19 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14088 Supplier BIOHIPPO Supplier No. BHE15212421
Rat PP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PP in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 8 19 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color