Learn More
Biomatik Corporation Rat Pregnane X Receptor (PXR) ELISA Kit
Supplier:  Biomatik Corporation EKU06756
Encompass_Preferred
Rat Pregnane X Receptor (PXR) ELISA Kit for detecting PXR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R1A7. Gene ID: 84385. Target Synonyms: NR1I2; SAR; PAR1; PAR2; PRR; BXR; ONR1; PAR; PARq; SXR; Nuclear Receptor Subfamily 1,Group I,Member 2; Steroid and xenobiotic receptor; Pregnane X receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6438