Learn More
Sino Biological Rat PRL2A1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80872R08H100
Encompass_Preferred
A DNA sequence encoding the rat Prl2a1 (NP_446243.1) (Met1-Cys228) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2120