Learn More
BIOHIPPO Rat Pro-ANP Microsample
SDP
Catalog No. 5000314642
Encompass
Rat Pro-ANP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Pro-ANP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 15 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat Pro-ANP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat Pro-ANP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat Pro-ANP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14642 Supplier BIOHIPPO Supplier No. BHE15210708
Rat Pro-ANP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Pro-ANP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 15 2 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat Pro-ANP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat Pro-ANP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat Pro-ANP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color