Learn More
BIOHIPPO Rat pro Microsample
SDP
Catalog No. 5000314598
Encompass
Rat pro Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of pro in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 16 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat pro-IL1b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat pro-IL1b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat pro-IL1b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14598 Supplier BIOHIPPO Supplier No. BHE15210699
Rat pro Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of pro in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 16 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat pro-IL1b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat pro-IL1b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat pro-IL1b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color