Learn More
American Research Products Inc Rat Pro opiomelanocortin POMC
Catalog No. 5000329410
Encompass
Rat Pro-opiomelanocortin (POMC) ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29410 Supplier American Research Products Inc Supplier No. CSBE13569R
Rat Pro-opiomelanocortin (POMC) ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich