Learn More
Biomatik Corporation Rat Procollagen I C-Terminal Propeptide (PICP) CLIA Kit
Supplier:  Biomatik Corporation EKU09955
Encompass_Preferred
Rat Procollagen I C-Terminal Propeptide (PICP) CLIA Kit for detecting PICP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.21-150ng/ml. Sensitivity: 0.09ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02454. Gene ID: 29393. Target Synonyms: P1CP; C-Propeptide Of Type I Procollagen; Procollagen I Carboxy Terminal Propeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8395