Learn More
American Research Products Inc Rat procollagen I N terminal p
Catalog No. 5000329153
Encompass
Rat procollagen N-terminal peptide P NP ELISA Kit from CUSABIO Detection Range 62 5 pg/ml-4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29153 Supplier American Research Products Inc Supplier No. CSBE12774R
Rat procollagen N-terminal peptide P NP ELISA Kit from CUSABIO Detection Range 62 5 pg/ml-4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich