Learn More
Biomatik Corporation Rat Procollagen III (PCIII) ELISA Kit
Supplier:  Biomatik Corporation EKU06784
Encompass_Preferred
Rat Procollagen III (PCIII) ELISA Kit for detecting PCIII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.5-160ng/ml. Sensitivity: 0.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13941. Gene ID: 84032. Target Synonyms: PCOL-III; PC-III; PC3; Type III Procollagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8807