Learn More
Crystal Chem Inc Rat Progesterone ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80558
Encompass
The Rat Progesterone ELISA is a highly sensitive assay that is used to quantify levels of rat progesterone in serum and plasma.Highlights: Kit uses sample size of only 25 µL. Dynamic range of 0.4 - 100 ng/mL. No extraction needed, ready to use reagents. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7729