Learn More
Cayman Chemical Rat Progesterone Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502947425
Encompass_Preferred
Rat PGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spinand-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Item Nos. R05001-32, R05001]
Catalog No. 50-294-7425 Supplier Cayman Chemical Supplier No. 3258196 WELL
Rat PGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spinand-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Item Nos. R05001-32, R05001]